missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein-arginine deiminase type-2 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein-arginine deiminase type-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8266-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_031391.2 | |
Protein-arginine deiminase type-2 Protein | |
Research Use Only | |
PADI2 | |
Recombinant Protein | |
Yeast | |
MLRERTVRLQ YGSRVEAVYV LGTYLWTDVY SAAPAGAQTF SLKHSEHVWV EVVRDGEAEE VATNGKQRWL LSPSTTLRVT MSQASTEASS DKVTVNYYDE EGSIPIDQAG LFLTAIEISL DVDADRDGVV EKNNPKKASW TWGPEGQGAI LLVNCDRETP WLPKEDCRDE KVYSKEDLKD MSQMILRTKG PDRLPAGYEI VLYISMSDSD KVGVFYVENP FFGQRYIHIL GRRKLYHVVK YTGGSAELLF FVEGLCFPDE GFSGLVSIHV SLLEYMAQDI PLTPIFTDTV IFRIAPWIMT PNILPPVSVF VCCMKDNYLF LKEVKNLVEK TNCELKVCFQ YLNRGDRWIQ DEIEFGYIEA PHKGFPVVLD SPRDGNLKDF PVKELLGPDF GYVTREPLFE SVTSLDSFGN LEVSPPVTVN GKTYPLGRIL IGSSFPLSGG RRMTKVVRDF LKAQQVQAPV ELYSDWLTVG HVDEFMSFVP IPGTKKFLLL MASTSACYKL FREKQKDGHG EAIMFKGLGG MSSKRITINK ILSNESLVQE NLYFQRCLDW NRDILKKELG LTEQDIIDLP ALFKMDEDHR ARAFFPNMVN MIVLDKDLGI PKPFGPQVEE ECCLEMHVRG LLEPLGLECT FIDDISAYHK FLGEVHCGTN VRRKPFTFKW WHMVP | |
Greater than 90% as determined by SDS-PAGE. |
11240 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |