missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein AATF Protein

A DNA sequence encoding the Homo sapiens (Human) Protein AATF, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8502-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16064981

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_036270.1
Protein AATF Protein
Research Use Only
AATF
Recombinant Protein
E. coli
PLALQLEQLL NPRPSEADPE ADPEEATAAR VIDRFDEGED GEGDFLVVGS IRKLASASLL DTDKRYCGKT TSRKAWNEDH WEQTLPGSSD EEISDEEGSG DEDSEGLGLE EYDEDDLGAA EEQECGDHRE SKKSRSHSAK TPGFSVQSIS DFEKFTKGMD DLGSSEEEED EESGMEEGDD AEDSQGESEE DRAGDRNSED DGVVMTFSSV KVSEEVEKGR AVKNQIALWD QLLEGRIKLQ KALLTTNQLP QPDVFPLFKD KGGPEFSSAL KNSHKALKAL LRSLVGLQE
Greater than 90% as determined by SDS-PAGE.
26574
500 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers