missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Proteasome subunit alpha type-1 Protein

A DNA sequence encoding the Homo sapiens (Human) Proteasome subunit alpha type-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6543-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16034050

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_002777.1
Proteasome subunit alpha type-1 Protein
Research Use Only
PSMA1
Recombinant Protein
E. coli
MFRNQYDNDV TVWSPQGRIH QIEYAMEAVK QGSATVGLKS KTHAVLVALK RAQSELAAHQ KKILHVDNHI GISIAGLTAD ARLLCNFMRQ ECLDSRFVFD RPLPVSRLVS LIGSKTQIPT QRYGRRPYGV GLLIAGYDDM GPHIFQTCPS ANYFDCRAMS IGARSQSART YLERHMSEFM ECNLNELVKH GLRALRETLP AEQDLTTKNV SIGIVGKDLE FTIYDDDDVS PFLEGLEERP QRKAQPAQPA D
Greater than 90% as determined by SDS-PAGE.
5682
500 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers