missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Proline-rich protein 11 Protein
A DNA sequence encoding the Homo sapiens (Human) Proline-rich protein 11, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7874-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_060774.2 | |
Proline-rich protein 11 Protein | |
Research Use Only | |
PRR11 | |
Recombinant Protein | |
E. coli | |
MPKFKQRRRK LKAKAERLFK KKEASHFQSK LITPPPPPPS PERVGISSID ISQSRSWLTS SWNFNFPNIR DAIKLWTNRV WSIYSWCQNC ITQSLEVLKD TIFPSRICHR ELYSVKQQFC ILESKLCKLQ EALKTISESS SCPSCGQTCH MSGKLTNVPA CVLITPGDSK AVLPPTLPQP ASHFPPPPPP PPLPPPPPPL APVLLRKPSL AKALQAGPLK KDGPMQITVK DLLTVKLKKT QSLDEKRKLI PSPKARNPLV TVSDLQHVTL KPNSKVLSTR VTNVLITPGK SQMDLRKLLR KVDVERSPGG TPLTNKENME TGTGLTPVMT QALRRKFQLA HPRSPTPTLP LSTSSFDEQN | |
Greater than 90% as determined by SDS-PAGE. |
55771 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |