missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PPIAL4A Protein
A DNA sequence encoding the Homo sapiens (Human) PPIAL4A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8269-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001137355.1 | |
PPIAL4A Protein | |
Research Use Only | |
PPIAL4A | |
Recombinant Protein | |
E. coli | |
MVNSVVFFDI TVDGKPLGRI SIKLFADKIL KTAENFRALS TGEKGFRYKG SCFHRIIPGF MCQGGDFTRH NGTGDKSIYG EKFDDENLIR KHTGSGILSM ANAGPNTNGS QFFICAAKTE WLDGKHVAFG KVKERVNIVE AMEHFGYRNS KTSKKITIAD CGQF | |
Greater than 90% as determined by SDS-PAGE. |
653505 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |