missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Polyadenylate-binding protein 1

A DNA sequence encoding the Homo sapiens (Human) Polyadenylate-binding protein 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6461-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16093460

  • 2180.00€ / 1mg

Estimated Shipment: 28-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_002559.2
Polyadenylate-binding protein 1
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
26986
Greater than 90% as determined by SDS-PAGE.
Research Use Only
PABPC1
Recombinant Protein
E. coli
MNPSAPSYPM ASLYVGDLHP DVTEAMLYEK FSPAGPILSI RVCRDMITRR SLGYAYVNFQ QPADAERALD TMNFDVIKGK PVRIMWSQRD PSLRKSGVGN IFIKNLDKSI DNKALYDTFS AFGNILSCKV VCDENGSKGY GFVHFETQEA AERAIEKMNG MLLNDRKVFV GRFKSRKERE AELGARAKEF TNVYIKNFGE DMDDERLKDL FGKFGPALSV KVMTDESGKS KGFGFVSFER HEDAQKAVDE MNGKELNGKQ IYVGRAQKKV ERQTELKRKF EQMKQDRITR YQGVNLYVKN LDDGIDDERL RKEFSPFGTI TSAKVMMEGG RSKGFGFVCF SSPEEATKAV TEMNGRIVAT KPLYVALAQR
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers