missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Polyadenylate-binding protein 1
A DNA sequence encoding the Homo sapiens (Human) Polyadenylate-binding protein 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6461-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002559.2 | |
Polyadenylate-binding protein 1 | |
Research Use Only | |
PABPC1 | |
Recombinant Protein | |
E. coli | |
MNPSAPSYPM ASLYVGDLHP DVTEAMLYEK FSPAGPILSI RVCRDMITRR SLGYAYVNFQ QPADAERALD TMNFDVIKGK PVRIMWSQRD PSLRKSGVGN IFIKNLDKSI DNKALYDTFS AFGNILSCKV VCDENGSKGY GFVHFETQEA AERAIEKMNG MLLNDRKVFV GRFKSRKERE AELGARAKEF TNVYIKNFGE DMDDERLKDL FGKFGPALSV KVMTDESGKS KGFGFVSFER HEDAQKAVDE MNGKELNGKQ IYVGRAQKKV ERQTELKRKF EQMKQDRITR YQGVNLYVKN LDDGIDDERL RKEFSPFGTI TSAKVMMEGG RSKGFGFVCF SSPEEATKAV TEMNGRIVAT KPLYVALAQR | |
Greater than 90% as determined by SDS-PAGE. |
26986 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |