missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Phytanoyl-CoA dioxygenase, peroxisomal Protein

A DNA sequence encoding the Homo sapiens (Human) Phytanoyl-CoA dioxygenase, peroxisomal, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6492-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16023690

  • 530.00€ / 100µg
Estimated Shipment: 25-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_001032626.1
Phytanoyl-CoA dioxygenase, peroxisomal Protein
Research Use Only
PHYH
Recombinant Protein
E. coli
SGTISSASFH PQQFQYTLDN NVLTLEQRKF YEENGFLVIK NLVPDADIQR FRNEFEKICR KEVKPLGLTV MRDVTISKSE YAPSEKMITK VQDFQEDKEL FRYCTLPEIL KYVECFTGPN IMAMHTMLIN KPPDSGKKTS RHPLHQDLHY FPFRPSDLIV CAWTAMEHIS RNNGCLVVLP GTHKGSLKPH DYPKWEGGVN KMFHGIQDYE ENKARVHLVM EKGDTVFFHP LLIHGSGQNK TQGFRKAISC HFASADCHYI DVKGTSQENI EKEVVGIAHK FFGAENSVNL KDIWMFRARL VKGERTNL
Greater than 80% as determined by SDS-PAGE.
5264
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers