missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PGDH / PHGDH Protein
A DNA sequence encoding the Homo sapiens (Human) PGDH / PHGDH, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8438-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006614.2 | |
PGDH / PHGDH Protein | |
Research Use Only | |
PHGDH | |
Recombinant Protein | |
E. coli | |
AFANLRKVLI SDSLDPCCRK ILQDGGLQVV EKQNLSKEEL IAELQDCEGL IVRSATKVTA DVINAAEKLQ VVGRAGTGVD NVDLEAATRK GILVMNTPNG NSLSAAELTC GMIMCLARQI PQATASMKDG KWERKKFMGT ELNGKTLGIL GLGRIGREVA TRMQSFGMKT IGYDPIISPE VSASFGVQQL PLEEIWPLCD FITVHTPLLP STTGLLNDNT FAQCKKGVRV VNCARGGIVD EGALLRALQS | |
Greater than 90% as determined by SDS-PAGE. |
26227 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |