missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PGD / Phosphogluconate dehydrogenase Protein
A DNA sequence encoding the Homo sapiens (Human) PGD / Phosphogluconate dehydrogenase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8351-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001291380.1 | |
PGD / Phosphogluconate dehydrogenase Protein | |
Research Use Only | |
PGD | |
Recombinant Protein | |
E. coli | |
ADIALIGLAV MGQNLILNMN DHGFVVCAFN RTVSKVDDFL ANEAKGTKVV GAQSLKEMVS KLKKPRRIIL LVKAGQAVDD FIEKLVPLLD TGDIIIDGGN SEYRDTTRRC RDLKAKGILF VGSGVSGGEE GARYGPSLMP GGNKEAWPHI KTIFQGIAAK VGTGEPCCDW VGDEGAGHFV KMVHNGIEYG DMQLICEAYH LMKDVLGMAQ DEMAQAFEDW NKTELDSFLI EITANILKFQ DTDGKHLLPK IRDSAGQKGT GKWTAISALE YGVPVTLIGE AVFARCLSSL KDERIQASKK LKGPQKFQFD GDKKSFLEDI RKALYASKII SYAQGFMLLR QAATEFGWTL NYGGIALMWR GGCIIRSVFL GKIKDAFDRN PELQNLLLDD FFKSAVENCQ DSWRRAVSTG VQAGIPMPCF TTALSFYDGY RHEMLPASLI QAQRDYFGAH TYELLAKPGQ FIHTNWTGHG GTVSSSSYNA | |
Greater than 90% as determined by SDS-PAGE. |
5226 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |