missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PGD / Phosphogluconate dehydrogenase Protein

A DNA sequence encoding the Homo sapiens (Human) PGD / Phosphogluconate dehydrogenase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8351-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16024171

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001291380.1
PGD / Phosphogluconate dehydrogenase Protein
Research Use Only
PGD
Recombinant Protein
E. coli
ADIALIGLAV MGQNLILNMN DHGFVVCAFN RTVSKVDDFL ANEAKGTKVV GAQSLKEMVS KLKKPRRIIL LVKAGQAVDD FIEKLVPLLD TGDIIIDGGN SEYRDTTRRC RDLKAKGILF VGSGVSGGEE GARYGPSLMP GGNKEAWPHI KTIFQGIAAK VGTGEPCCDW VGDEGAGHFV KMVHNGIEYG DMQLICEAYH LMKDVLGMAQ DEMAQAFEDW NKTELDSFLI EITANILKFQ DTDGKHLLPK IRDSAGQKGT GKWTAISALE YGVPVTLIGE AVFARCLSSL KDERIQASKK LKGPQKFQFD GDKKSFLEDI RKALYASKII SYAQGFMLLR QAATEFGWTL NYGGIALMWR GGCIIRSVFL GKIKDAFDRN PELQNLLLDD FFKSAVENCQ DSWRRAVSTG VQAGIPMPCF TTALSFYDGY RHEMLPASLI QAQRDYFGAH TYELLAKPGQ FIHTNWTGHG GTVSSSSYNA
Greater than 90% as determined by SDS-PAGE.
5226
200 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers