missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Perilipin-4 Protein
A DNA sequence encoding the Homo sapiens (Human) Perilipin-4, was expressed in the hosts and tags indicated.
287.00€ - 3175.00€
Specifications
Accession Number | NP_001073869.1 |
---|---|
Gene ID (Entrez) | 729359 |
Name | Perilipin-4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16001161
|
enQuireBio™
QP7848-YE-100UG |
100 μg |
920.00€
100µg |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
16041161
|
enQuireBio™
QP7848-YE-500UG |
500 μg |
2650.00€
500µg |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
16011161
|
enQuireBio™
QP7848-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16031161
|
enQuireBio™
QP7848-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16051161
|
enQuireBio™
QP7848-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16021161
|
enQuireBio™
QP7848-YE-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001073869.1 | |
Perilipin-4 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
729359 | |
Research Use Only | |
PLIN4 | |
Recombinant Protein | |
Yeast | |
GVDTSKTVLT GTKDTVCSGV TGAMNVAKGT IQTGVDTTKT VLTGTKNTVC SGVTGAVNLA KEAIQGGLDT TKSMVMGTKD TMSTGLTGAA NVAKGAMQTG LNTTQNIATG T | |
Greater than 90% as determined by SDS-PAGE. |