missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Perforin-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Perforin-1, was expressed in the hosts and tags indicated.
234.00€ - 2385.00€
Specifications
Accession Number | NP_001076585.1 |
---|---|
Gene ID (Entrez) | 5551 |
Name | Perforin-1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16013920
|
enQuireBio™
QP6525-EC-10UG |
10 μg |
251.00€
10µg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16003920
|
enQuireBio™
QP6525-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
16023920
|
enQuireBio™
QP6525-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
16033920
|
enQuireBio™
QP6525-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16043920
|
enQuireBio™
QP6525-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16053920
|
enQuireBio™
QP6525-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001076585.1 | |
Perforin-1 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
5551 | |
Research Use Only | |
Prf1 | |
Recombinant Protein | |
E. coli | |
PCHTAARSEC KRSHKFVPGA WLAGEGVDVT SLRRSGSFPV DTQRFLRPDG TCTLCENALQ EGTLQRLPLA LTNWRAQGSG CQRHVTRAKV SSTEAVARDA ARSIRNDWKV GLDVTPKPTS NVHVSVAGSH SQAANFAAQK THQDQYSFST DTVECRFYSF HVVHTPPLHP DFKRALGDLP HHFNASTQPA YLRLISNYGT HFIRAVELGG RISALTALRT CELALEGLTD NEVEDCLTVE AQVNIGIHGS ISAEAKACEE KKKKHKMTAS FHQTYRERHS EVVGGHHTSI NDLLFGIQAG PEQYSAWVNS LPGSPGLVDY TLEPLHVLLD SQDPRREALR RALSQYLTDR ARWRDCSRPC PPGRQKSPRD PCQCVCHGSA VTTQDCCPRQ RGLAQLEVTF IQAWGLWGDW FTATDAYVKL FFGGQELRTS TVWDNNNPIW SVRLDFGDVL LATGGPLRLQ VWDQDSGRDD DLLGTCDQAP KSGSHEVRCN LNHGHLKFRY HARCLPHLGG GTCLDYVPQM LLGEPPGNRS GAVW | |
Greater than 80% as determined by SDS-PAGE. |