missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PC4 and SFRS1-interacting Protein
A DNA sequence encoding the Homo sapiens (Human) PC4 and SFRS1-interacting protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6542-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001121689.1 | |
PC4 and SFRS1-interacting Protein | |
Research Use Only | |
PSIP1 | |
Recombinant Protein | |
E. coli | |
MTRDFKPGDL IFAKMKGYPH WPARVDEVPD GAVKPPTNKL PIFFFGTHET AFLGPKDIFP YSENKEKYGK PNKRKGFNEG LWEIDNNPKV KFSSQQAATK QSNASSDVEV EEKETSVSKE DTDHEEKASN EDVTKAVDIT TPKAARRGRK RKAEKQVETE EAGVVTTATA SVNLKVSPKR GRPAATEVKI PKPRGRPKMV KQPCPSESDI ITEEDKSKKK GQEEKQPKKQ PKKDEEGQKE EDKPRKEPDK KEGKKEVESK RKNLAKTGVT STSDSEEEGD DQEGEKKRKG GRNFQTAHRR NMLKGQHEKE AADRKRKQEE QMETEQQNKD EGKKPEVKKV EKKRETSMDS RLQRIHAEIK NSLKIDNLDV NRCIEALDEL ASLQVTMQQA QKHTEMITTL KKIRRFKVSQ VIMEKSTMLY NKFKNMFLVG EGDSVITQVL NKSLAEQRQH EEANKTKDQG KKGPNKKLEK EQTGSKTLNG GSDAQDGNQP QHNGESNEDS KDNHEASTKK KPSSEERETE ISLKDSTLDN | |
Greater than 80% as determined by SDS-PAGE. |
11168 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |