missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PC4 and SFRS1-interacting Protein

A DNA sequence encoding the Homo sapiens (Human) PC4 and SFRS1-interacting protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6542-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16064040

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001121689.1
PC4 and SFRS1-interacting Protein
Research Use Only
PSIP1
Recombinant Protein
E. coli
MTRDFKPGDL IFAKMKGYPH WPARVDEVPD GAVKPPTNKL PIFFFGTHET AFLGPKDIFP YSENKEKYGK PNKRKGFNEG LWEIDNNPKV KFSSQQAATK QSNASSDVEV EEKETSVSKE DTDHEEKASN EDVTKAVDIT TPKAARRGRK RKAEKQVETE EAGVVTTATA SVNLKVSPKR GRPAATEVKI PKPRGRPKMV KQPCPSESDI ITEEDKSKKK GQEEKQPKKQ PKKDEEGQKE EDKPRKEPDK KEGKKEVESK RKNLAKTGVT STSDSEEEGD DQEGEKKRKG GRNFQTAHRR NMLKGQHEKE AADRKRKQEE QMETEQQNKD EGKKPEVKKV EKKRETSMDS RLQRIHAEIK NSLKIDNLDV NRCIEALDEL ASLQVTMQQA QKHTEMITTL KKIRRFKVSQ VIMEKSTMLY NKFKNMFLVG EGDSVITQVL NKSLAEQRQH EEANKTKDQG KKGPNKKLEK EQTGSKTLNG GSDAQDGNQP QHNGESNEDS KDNHEASTKK KPSSEERETE ISLKDSTLDN
Greater than 80% as determined by SDS-PAGE.
11168
200 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers