missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PA28A / PSME1 Protein
A DNA sequence encoding the Homo sapiens (Human) PA28A / PSME1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6547-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001268457.1 | |
PA28A / PSME1 Protein | |
Research Use Only | |
PSME1 | |
Recombinant Protein | |
E. coli | |
MAMLRVQPEA QAKVDVFRED LCTKTENLLG SYFPKKISEL DAFLKEPALN EANLSNLKAP LDIPVPDPVK EKEKEERKKQ QEKEDKDEKK KGEDEDKGPP CGPVNCNEKI VVLLQRLKPE IKDVIEQLNL VTTWLQLQIP RIEDGNNFGV AVQEKVFELM TSLHTKLEGF HTQISKYFSE RGDAVTKAAK QPHVGDYRQL VHELDEAEYR DIRLMVMEIR NAYAVLYDII LKNFEKLKKP RGETKGMIY | |
Greater than 80% as determined by SDS-PAGE. |
5720 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |