missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NR2C2 Protein
A DNA sequence encoding the Homo sapiens (Human) NR2C2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8855-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001334271.1 | |
NR2C2 Protein | |
Research Use Only | |
NR2C2 | |
Recombinant Protein | |
E. coli | |
SPSPRIQIIS TDSAVASPQR IQIVTDQQTG QKIQIVTAVD ASGSSKQQFI LTSPDGAGTG KVILASPETS SAKQLIFTTS DNLVPGRIQI VTDSASVERL LGKADVQRPQ VVEYCVVCGD KASGRHYGAV SCEGCKGFFK RSVRKNLTYS CRSSQDCIIN KHHRNRCQFC RLKKCLEMGM KMESVQSERK PFDVQREKPS NCAASTEKIY IRKDLRSPLI ATPTFVADKD GARQTGLLDP GMLVNIQQPL IREDGTVLLA ADSKAETSQG ALGTLANVVT SLANLSESLN NGDASEMQPE DQSASEITRA FDTLAKALNT TDSASPPSLA DGIDASGGGS IHVISRDQST PIIEVEGPLL SDTHVTFKLT MPSPMPEYLN VHYICESASR LLFLSMHWAR SIPAFQALGQ DCNTSLVRAC WNELFTLGLA QCAQVMSLST ILAAIVNHLQ NSIQEDKLSG DRIKQVMEHI WKLQEFCNSM AKLDIDGYEY AYLKAIVLFS PDHPGLTGTS QIEKFQEKAQ MELQDYVQKT YSEDTYRLAR ILVRLPALRL MSSNITEELF FTGLIGNVSI DSIIPYILKM ETAEYNGQIT | |
Greater than 90% as determined by SDS-PAGE. |
22026 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |