missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NME1 / NDKA Protein
A DNA sequence encoding the Homo sapiens (Human) NME1 / NDKA, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8426-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000260.1 | |
NME1 / NDKA Protein | |
Research Use Only | |
NME1 | |
Recombinant Protein | |
E. coli | |
ANCERTFIAI KPDGVQRGLV GEIIKRFEQK GFRLVGLKFM QASEDLLKEH YVDLKDRPFF AGLVKYMHSG PVVAMVWEGL NVVKTGRVML GETNPADSKP GTIRGDFCIQ VGRNIIHGSD SVESAEKEIG LWFHPEELVD YTSCAQNWIY E | |
Greater than 90% as determined by SDS-PAGE. |
4830 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |