missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Nephrocystin-1 Protein

A DNA sequence encoding the Homo sapiens (Human) Nephrocystin-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6428-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16033240

  • 530.00€ / 100µg
Estimated Shipment: 24-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_000263.2
Nephrocystin-1 Protein
Research Use Only
NPHP1
Recombinant Protein
E. coli
MLARRQRDPL QALRRRNQEL KQQVDSLLSE SQLKEALEPN KRQHIYQRCI QLKQAIDENK NALQKLSKAD ESAPVANYNQ RKEEEHTLLD KLTQQLQGLA VTISRENIT
Greater than 90% as determined by SDS-PAGE.
4867
100 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers