missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NDUFS5 Protein

A DNA sequence encoding the Homo sapiens (Human) NDUFS5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8422-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16024531

  • 530.00€ / 100µg
Estimated Shipment: 25-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_001171908.1
NDUFS5 Protein
Research Use Only
NDUFS5
Recombinant Protein
E. coli
PFLDIQKRFG LNIDRWLTIQ SGEQPYKMAG RCHAFEKEWI ECAHGIGYTR AEKECKIEYD DFVECLLRQK TMRRAGTIRK QRDKLIKEGK YTPPPHHIGK GEPRP
Greater than 90% as determined by SDS-PAGE.
4725
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers