missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Myosin-6 Protein
A DNA sequence encoding the Homo sapiens (Human) Myosin-6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9299-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002462.2 | |
Myosin-6 Protein | |
Research Use Only | |
MYH6 | |
Recombinant Protein | |
Yeast | |
NAYQYMLTDR ENQSILITGE SGAGKTVNTK RVIQYFASIA AIGDRGKKDN ANANKGTLED QIIQANPALE AFGNAKTVRN DNSSRFGKFI RIHFGATGKL ASADIETYLL EKSRVIFQLK AERNYHIFYQ ILSNKKPELL DMLLVTNNPY DYAFVSQGEV SVASIDDSEE LMATDSAFDV LGFTSEEKAG VYKLTGAIMH YGNMKFKQKQ REEQAEPDGT EDADKSAYLM GLNSADLLKG LCHPRVKVGN EYVTKGQSVQ QVYYSIGALA KAVYEKMFNW MVTRINATLE TKQPRQYFIG VLDIAGFEIF DFNSFEQLCI NFTNEKLQQF FNHHMFVLEQ EEYKKEGIEW TFIDFGMDLQ ACIDLIEKPM GIMSILEEEC MFPKATDMTF KAKLYDNHLG KSNNFQKPRN IKGKQEAHFS LIHYAGTVDY NILGWLEKNK DPLNETVVAL YQKSSLKLMA TLFSSYATAD TGDSGKSKGG KKKGSSFQTV SALHRENLNK LMTNLRTTHP HFVRCIIPNE RKAPGVMDNP LVMHQLRCNG VLEGIRICRK GFPNRILYGD FRQRYRILNP VAIPEGQFID SRKGTEKLLS SLDIDHNQYK FGHTKVFFKA GLLGLLEEMR DERLSRIITR MQAQARGQLM RIEFKKIVER RDALLVI | |
Greater than 90% as determined by SDS-PAGE. |
4624 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |