missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Myelin transcription factor 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Myelin transcription factor 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8849-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16036821

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_004526.1
Myelin transcription factor 1 Protein
Research Use Only
MYT1
Recombinant Protein
E. coli
GLGHISGKYA SHRSASGCPL AARRQKEGSL NGSSFSWKSL KNEGPTCPTP GCDGSGHANG SFLTHRSLSG CPRATFAGKK GKLSGDEVLS PKFKTSDVLE NDEEIKQLNQ EIRDLNESNS EMEAAMVQLQ SQISSMEKNL KNIEEENKLI EEQNEALFLE LSGLSQALIQ SLANIRLPHM EPICEQNFDA YVSTLTDMYS NQ
Greater than 90% as determined by SDS-PAGE.
4661
500 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers