missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Myelin transcription factor 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Myelin transcription factor 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8849-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16016821

  • 2385.00€ / 1mg
Estimated Shipment: 28-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_004526.1
Myelin transcription factor 1 Protein
Research Use Only
MYT1
Recombinant Protein
E. coli
GLGHISGKYA SHRSASGCPL AARRQKEGSL NGSSFSWKSL KNEGPTCPTP GCDGSGHANG SFLTHRSLSG CPRATFAGKK GKLSGDEVLS PKFKTSDVLE NDEEIKQLNQ EIRDLNESNS EMEAAMVQLQ SQISSMEKNL KNIEEENKLI EEQNEALFLE LSGLSQALIQ SLANIRLPHM EPICEQNFDA YVSTLTDMYS NQ
Greater than 90% as determined by SDS-PAGE.
4661
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers