missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Mitochondrial import receptor subunit TOM34 Protein

A DNA sequence encoding the Homo sapiens (Human) Mitochondrial import receptor subunit TOM34, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7602-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16069680

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_006800.2
Mitochondrial import receptor subunit TOM34 Protein
Research Use Only
TOMM34
Recombinant Protein
E. coli
MAPKFPDSVE ELRAAGNESF RNGQYAEASA LYGRALRVLQ AQGSSDPEEE SVLYSNRAAC HLKDGNCRDC IKDCTSALAL VPFSIKPLLR RASAYEALEK YPMAYVDYKT VLQIDDNVTS AVEGINRMTR ALMDSLGPEW RLKLPSIPLV PVSAQKRWNS LPSENHKEMA KSKSKETTAT KNRVPSAGDV EKARVLKEEG NELVKKGNHK KAIEKYSESL LCSNLESATY SNRALCYLVL KQYTEAVKDC TEALKLDGKN VKAFYRRAQA HKALKDYKSS FADISNLLQI EPRNGPAQKL RQEVKQNLH
Greater than 80% as determined by SDS-PAGE.
10953
500 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers