missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Mitochondrial fission process protein 1

A DNA sequence encoding the Homo sapiens (Human) Mitochondrial fission process protein 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8083-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16072561

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001003704.1
Mitochondrial fission process protein 1
Research Use Only
MTFP1
Recombinant Protein
E. coli
MSEPQPRGAE RDLYRDTWVR YLGYANEVGE AFRSLVPAAV VWLSYGVASS YVLADAIDKG KKAGEVPSPE AGRSARVTVA VVDTFVWQAL ASVAIPGFTI NRVCAASLYV LGTATRWPLA VRKWTTTALG LLTIPIIIHP IDRSVDFLLD SSLRKLYPTV GKPSSS
Greater than 80% as determined by SDS-PAGE.
51537
200 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers