missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MIP-3 beta / CCL19 Protein
A DNA sequence encoding the Homo sapiens (Human) MIP-3 beta / CCL19, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8024-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006265.1 | |
MIP-3 beta / CCL19 Protein | |
Research Use Only | |
CCL19 | |
Recombinant Protein | |
E. coli | |
GTNDAEDCCL SVTQKPIPGY IVRNFHYLLI KDGCRVPAVV FTTLRGRQLC APPDQPWVER IIQRLQRTSA KMKRRSS | |
Greater than 90% as determined by SDS-PAGE. |
6363 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |