missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MIP-3 beta / CCL19 Protein

A DNA sequence encoding the Homo sapiens (Human) MIP-3 beta / CCL19, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8024-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16062221

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_006265.1
MIP-3 beta / CCL19 Protein
Research Use Only
CCL19
Recombinant Protein
E. coli
GTNDAEDCCL SVTQKPIPGY IVRNFHYLLI KDGCRVPAVV FTTLRGRQLC APPDQPWVER IIQRLQRTSA KMKRRSS
Greater than 90% as determined by SDS-PAGE.
6363
500 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers