missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MFG-E8 / lactadherin Protein

A DNA sequence encoding the Homo sapiens (Human) MFG-E8 / lactadherin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6356-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16062710

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001108086.1
MFG-E8 / lactadherin Protein
Research Use Only
MFGE8
Recombinant Protein
E. coli
LDICSKNPCH NGGLCEEISQ EVRGDVFPSY TCTCLKGYAG NHCETKCVEP LGMENGNIAN SQIAASSVRV TFLGLQHWVP ELARLNRAGM VNAWTPSSND DNPWIQVNLL RRMWVTGVVT QGASRLASHE YLKAFKVAYS LNGHEFDFIH DVNKKHKEFV GNWNKNAVHV NLFETPVEAQ YVRLYPTSCH TACTLRFELL GCELNGCANP LGLKNNSIPD KQITASSSYK TWGLHLFSWN PSYARLDKQG NFNAWVAGSY GNDQWLQVDL GSSKEVTGII TQGARNFGSV QFVASYKVAY SNDSANWTEY QDPRTGSSKI FPGNWDNHSH KKNLFETPIL ARYVRILPVA WHNRIALRLE LLGC
Greater than 90% as determined by SDS-PAGE.
4240
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers