missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Metallothionein-3 Protein

A DNA sequence encoding the Homo sapiens (Human) Metallothionein-3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6390-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16052970

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_005945.1
Metallothionein-3 Protein
Research Use Only
MT3
Recombinant Protein
E. coli
MDPETCPCPS GGSCTCADSC KCEGCKCTSC KKSCCSCCPA ECEKCAKDCV CKGGEAAEAE AEKCSCCQ
Greater than 90% as determined by SDS-PAGE.
4504
50 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers