missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Metallothionein-3 Protein
A DNA sequence encoding the Homo sapiens (Human) Metallothionein-3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6390-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_005945.1 | |
Metallothionein-3 Protein | |
Research Use Only | |
MT3 | |
Recombinant Protein | |
E. coli | |
MDPETCPCPS GGSCTCADSC KCEGCKCTSC KKSCCSCCPA ECEKCAKDCV CKGGEAAEAE AEKCSCCQ | |
Greater than 90% as determined by SDS-PAGE. |
4504 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |