missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Malate dehydrogenase, cytoplasmic Protein

A DNA sequence encoding the Homo sapiens (Human) Malate dehydrogenase, cytoplasmic, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8485-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16054881

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001186040.1
Malate dehydrogenase, cytoplasmic Protein
Research Use Only
MDHA
Recombinant Protein
E. coli
SEPIRVLVTG AAGQIAYSLL YSIGNGSVFG KDQPIILVLL DITPMMGVLD GVLMELQDCA LPLLKDVIAT DKEDVAFKDL DVAILVGSMP RREGMERKDL LKANVKIFKS QGAALDKYAK KSVKVIVVGN PANTNCLTAS KSAPSIPKEN FSCLTRLDHN RAKAQIALKL GVTANDVKNV IIWGNHSSTQ YPDVNHAKVK LQGKEVGVYE ALKDDSWLKG EFVTTVQQRG AAVIKARKLS SAMSAAKAIC DHVRDIWFGT PEGEFVSMGV ISDGNSYGVP DDLLYSFPVV IKNKTWKFVE GLPINDFSRE KMDLTAKELT EEKESAFEFL SS
Greater than 90% as determined by SDS-PAGE.
4190
50 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers