missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Malate dehydrogenase, cytoplasmic Protein
A DNA sequence encoding the Homo sapiens (Human) Malate dehydrogenase, cytoplasmic, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8485-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001186040.1 | |
Malate dehydrogenase, cytoplasmic Protein | |
Research Use Only | |
MDHA | |
Recombinant Protein | |
E. coli | |
SEPIRVLVTG AAGQIAYSLL YSIGNGSVFG KDQPIILVLL DITPMMGVLD GVLMELQDCA LPLLKDVIAT DKEDVAFKDL DVAILVGSMP RREGMERKDL LKANVKIFKS QGAALDKYAK KSVKVIVVGN PANTNCLTAS KSAPSIPKEN FSCLTRLDHN RAKAQIALKL GVTANDVKNV IIWGNHSSTQ YPDVNHAKVK LQGKEVGVYE ALKDDSWLKG EFVTTVQQRG AAVIKARKLS SAMSAAKAIC DHVRDIWFGT PEGEFVSMGV ISDGNSYGVP DDLLYSFPVV IKNKTWKFVE GLPINDFSRE KMDLTAKELT EEKESAFEFL SS | |
Greater than 90% as determined by SDS-PAGE. |
4190 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |