missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ly6 / PLAUR domain-containing protein 1

A DNA sequence encoding the Homo sapiens (Human) Ly6/PLAUR domain-containing protein 6, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7780-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16030751

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001182614.1
Ly6 / PLAUR domain-containing protein 1
Research Use Only
LYPD6
Recombinant Protein
E. coli
AQSRDFTVKD IIYLHPSTTP YPGGFKCFTC EKAADNYECN RWAPDIYCPR ETRYCYTQHT MEVTGNSISV TKRCVPLEEC LSTGCRDSEH EGHKVCTSCC EGNICNLPLP RNETDATFAT TSPINQTNGH PRCMSVIVSC LWLWLGLML
Greater than 90% as determined by SDS-PAGE.
130574
10 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers