missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LHPP Protein
A DNA sequence encoding the Homo sapiens (Human) LHPP, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8086-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001161352.1 | |
LHPP Protein | |
Research Use Only | |
LHPP | |
Recombinant Protein | |
E. coli | |
MAPWGKRLAG VRGVLLDISG VLYDSGAGGG TAIAGSVEAV ARLKRSRLKV RFCTNESQKS RAELVGQLQR LGFDISEQEV TAPAPAACQI LKEQGLRPYL LIHDGVRSEF DQIDTSNPNC VVIADAGESF SYQNMNNAFQ VLMELEKPVL ISLGKGRYYK ETSGLMLDVG PYMKALEYAC GIKAEVVGKP SPEFFKSALQ AIGVEAHQAV MIGDDIVGDV GGAQRCGMRA LQVRTGKFRP SDEHHPEVKA DGYVDNLAEA VDLLLQHADK | |
Greater than 90% as determined by SDS-PAGE. |
64077 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |