missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Kita-kyushu lung cancer antigen 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Kita-kyushu lung cancer antigen 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7682-ye-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16030161

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001017978.1
Kita-kyushu lung cancer antigen 1 Protein
Research Use Only
CT83
Recombinant Protein
Yeast
MNFYLLLASS ILCALIVFWK YRRFQRNTGE MSSNSTALAL VRPSSSGLIN SNTDNNLAVY DLSRDILNNF PHSIARQKRI LVNLSMVENK LVELEHTLLS KGFRGASPHR KST
Greater than 80% as determined by SDS-PAGE.
203413
10 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers