missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Kinesin light chain 3 Protein
A DNA sequence encoding the Homo sapiens (Human) Kinesin light chain 3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6265-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_803136.2 | |
Kinesin light chain 3 Protein | |
Research Use Only | |
KLC3 | |
Recombinant Protein | |
Yeast | |
MSVQVAAPGS AGLGPERLSP EELVRQTRQV VQGLEALRAE HHGLAGHLAE ALAGQGPAAG LEMLEEKQQV VSHSLEAIEL GLGEAQVLLA LSAHVGALEA EKQRLRSQAR RLAQENVWLR EELEETQRRL RASEESVAQL EEEKRHLEFL GQLRQYDPPA ESQQSESPPR RDSLASLFPS EEEERKGPEA AGAAAAQQGG YEIPARLRTL HNLVIQYAGQ GRYEVAVPLC RQALEDLERS SGHCHPDVAT MLNILALVYR DQNKYKEATD LLHDALQIRE QTLGPEHPAV AATLNNLAVL YGKRGRYREA EPLCQRALEI REKVLGADHP DVAKQLNNLA LLCQNQGKFE DVERHYARAL SIYEALGGPH DPNVAKTKNN LASAYLKQNK YQQAEELYKE ILHKEDLPAP LGAPNTGTAG DAEQALRRSS SLSKIRESIR RGSEKLVSRL RGEAAAGAAG MKRAMSLNTL NVDAPRAPGT QFPSWHLDKA PRTLSASTQD LSPH | |
Greater than 90% as determined by SDS-PAGE. |
147700 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |