missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Kidney mitochondrial carrier protein 1

A DNA sequence encoding the Homo sapiens (Human) Kidney mitochondrial carrier protein 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7723-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16040421

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001010875.1
Kidney mitochondrial carrier protein 1
Research Use Only
SLC25A30
Recombinant Protein
E. coli
MSALNWKPFV YGGLASITAE CGTFPIDLTK TRLQIQGQTN DAKFKEIRYR GMLHALVRIG REEGLKALYS GIAPAMLRQA SYGTIKIGTY QSLKRLFIER PEDETLPINV ICGILSGVIS STIANPTDVL KIRMQAQSNT IQGGMIGNFM NIYQQEGTRG LWKGVSLTAQ RAAIVVGVEL PVYDITKKHL ILSGLMGDTV YTHFLSSFTC GLAGALASNP VDVVRTRMMN QRVLRDGRCS GYTGTLDCLL QTWKNEGFFA LYKGFWPNWL RLGPWNIIFF VTYEQLKKLD L
Greater than 90% as determined by SDS-PAGE.
253512
10 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers