missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Josephin-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Josephin-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7617-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_055691.1 | |
Josephin-1 Protein | |
Research Use Only | |
JOSD1 | |
Recombinant Protein | |
E. coli | |
MSCVPWKGDK AKSESLELPQ AAPPQIYHEK QRRELCALHA LNNVFQDSNA FTRDTLQEIF QRLSPNTMVT PHKKSMLGNG NYDVNVIMAA LQTKGYEAVW WDKRRDVGVI ALTNVMGFIM NLPSSLCWGP LKLPLKRQHW ICVREVGGAY YNLDSKLKMP EWIGGESELR KFLKHHLRGK NCELLLVVPE EVEAHQSWRT DV | |
Greater than 90% as determined by SDS-PAGE. |
9929 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |