missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Intraflagellar transport protein 27 homolog Protein
A DNA sequence encoding the Homo sapiens (Human) Intraflagellar transport protein 27 homolog, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_001171172.1 |
---|---|
Gene ID (Entrez) | 11020 |
Name | Intraflagellar transport protein 27 homolog Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16022821
|
enQuireBio™
QP8131-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 27-06-2024
Log in to see stock. |
|||||
16042821
|
enQuireBio™
QP8131-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 27-06-2024
Log in to see stock. |
|||||
16032821
|
enQuireBio™
QP8131-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16052821
|
enQuireBio™
QP8131-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16062821
|
enQuireBio™
QP8131-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16072821
|
enQuireBio™
QP8131-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001171172.1 | |
Intraflagellar transport protein 27 homolog Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
11020 | |
Research Use Only | |
IFT27 | |
Recombinant Protein | |
E. coli | |
MVKLAAKCIL AGDPAVGKTA LAQIFRSDGA HFQKSYTLTT GMDLVVKTVP VPDTGDSVEL FIFDSAGKEL FSEMLDKLWE SPNVLCLVYD VTNEESFNNC SKWLEKARSQ APGISLPGVL VGNKTDLAGR RAVDSAEARA WALGQGLECF ETSVKEMENF EAPFHCLAKQ FHQLYREKVE VFRALA | |
Greater than 90% as determined by SDS-PAGE. |