Learn More
enQuireBio™ Recombinant Human Intraflagellar transport protein 22 homolog Protein
A DNA sequence encoding the Homo sapiens (Human) Intraflagellar transport protein 22 homolog, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP8067-ec-1mg
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16062471
  • 2385.00€ / 1mg
Estimated Shipment: 25-06-2024
Specifications
NP_001124292.1
Intraflagellar transport protein 22 homolog Protein
Research Use Only
RABL5
Recombinant Protein
E. coli
MLKAKILFVG PCESGKTVLA NFLTESSDIT EYSPTQGVRI LEFENPHVTS NNKGTGCEFE LWDCGGDAKF ESCWPALMKD AHGVVIVFNA DIPSHRKEME MWYSCFVQQP SLQDTQCMLI AHHKPGSGDD KGSLSLSPPL NKLKLVHSNL EDDPEEIRME FIKYLKSIIN SMSESRDREE MSIMT
Greater than 90% as determined by SDS-PAGE.
64792
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol