missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Interferon alpha-inducible protein 1
A DNA sequence encoding the Homo sapiens (Human) Interferon alpha-inducible protein 6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9195-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_002029.3 | |
Interferon alpha-inducible protein 1 | |
Research Use Only | |
IFI6 | |
Recombinant Protein | |
Yeast | |
GKKKCSESSD SGSGFWKALT FMAVGGGLAV AGLPALGFTG AGIAANSVAA SLMSWSAILN GGGVPAGGLV ATLQSLGAGG SSVVIGNIGA LMGYATHKYL DSEEDEE | |
Greater than 90% as determined by SDS-PAGE. |
2537 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |