missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Integrin-linked protein kinase Protein
A DNA sequence encoding the Homo sapiens (Human) Integrin-linked protein kinase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8343-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001014794.1 | |
Integrin-linked protein kinase Protein | |
Research Use Only | |
ILK1 | |
Recombinant Protein | |
E. coli | |
MDDIFTQCRE GNAVAVRLWL DNTENDLNQG DDHGFSPLHW ACREGRSAVV EMLIMRGARI NVMNRGDDTP LHLAASHGHR DIVQKLLQYK ADINAVNEHG NVPLHYACFW GQDQVAEDLV ANGALVSICN KYGEMPVDKA KAPLRELLRE RAEKMGQNLN RIPYKDTFWK GTTRTRPRNG TLNKHSGIDF KQLNFLTKLN ENHSGELWKG RWQGNDIVVK VLKVRDWS | |
Greater than 90% as determined by SDS-PAGE. |
3611 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |