missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Integrin-linked protein kinase Protein

A DNA sequence encoding the Homo sapiens (Human) Integrin-linked protein kinase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8343-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16004131

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001014794.1
Integrin-linked protein kinase Protein
Research Use Only
ILK1
Recombinant Protein
E. coli
MDDIFTQCRE GNAVAVRLWL DNTENDLNQG DDHGFSPLHW ACREGRSAVV EMLIMRGARI NVMNRGDDTP LHLAASHGHR DIVQKLLQYK ADINAVNEHG NVPLHYACFW GQDQVAEDLV ANGALVSICN KYGEMPVDKA KAPLRELLRE RAEKMGQNLN RIPYKDTFWK GTTRTRPRNG TLNKHSGIDF KQLNFLTKLN ENHSGELWKG RWQGNDIVVK VLKVRDWS
Greater than 90% as determined by SDS-PAGE.
3611
200 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers