missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Integrin alpha-V Protein
A DNA sequence encoding the Homo sapiens (Human) Integrin alpha-V, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9224-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001138471.1 | |
Integrin alpha-V Protein | |
Research Use Only | |
ITGAV | |
Recombinant Protein | |
Yeast | |
DLALSEGDIH TLGCGVAQCL KIVCQVGRLD RGKSAILYVK SLLWTETFMN KENQNHSYSL KSSASFNVIE FPYKNLPIED ITNSTLVTTN VTWGIQPAPM PVPVWVIILA VLAGLLLLAV LVFVMYRMGF FKRVRPPQEE QEREQLQPHE NGEGNSET | |
Greater than 90% as determined by SDS-PAGE. |
3685 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |