Learn More
R&D Systems™ Recombinant Human Integrin alpha V beta 1 Protein CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 6579-AVB-050
Description
The Recombinant Human Integrin alpha V beta 1 Protein is derived from CHO. The Recombinant Human Integrin alpha V beta 1 Protein has been validated for the following applications: Bioactivity.Specifications
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. | |
| 115 kDa ( alpha V subunit) and 86 kDa ( beta 1 subunit) | |
| Chinese Hamster Ovary cell line,CHO-derived human Integrin alpha V beta 1 protein, (N-terminus) Human Integrin alpha V (Phe31-Val992) Accession &Num; NP_002201 GGGSGGGSGGGS Acidic Tail HHHHHH Human Integrin beta 1 (Gln21-Asp728) Accession &Num; P05556 GGGSGGGSGGGS Basic Tail (C-terminus) | |
| <1.0 EU per 1μg of the protein by the LAL method. | |
| Reconstitute at 500μg/mL in water. |
| 3685 | |
| 50μg | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
| Unconjugated |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.