missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IGF-2 / IGF-II Protein
A DNA sequence encoding the Homo sapiens (Human) IGF-2 / IGF-II, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8601-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000603.1 | |
IGF-2 / IGF-II Protein | |
Research Use Only | |
IGF2 | |
Recombinant Protein | |
E. coli | |
ETLCGGELVD TLQFVCGDRG FYFSRPASRV SRRSRGIVEE CCFRSCDLAL LETYCATPAK SERDVSTPPT VLPDNFPRYP VGKFFQYDTW KQSTQRLRRG LPALLRARRG HVLAKELEAF REAKRHRPLI ALPTQDPAHG GAPPEMASNR K | |
Greater than 90% as determined by SDS-PAGE. |
3481 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |