missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IFNG / Interferon Gamma Protein

A DNA sequence encoding the Homo sapiens (Human) IFNG / Interferon Gamma Protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8548-ye-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16045221

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_000610.2
IFNG / Interferon Gamma Protein
Research Use Only
IFNG
Recombinant Protein
Yeast
QDPYVKEAEN LKKYFNAGHS DVADNGTLFL GILKNWKEES DRKIMQSQIV SFYFKLFKNF KDDQSIQKSV ETIKEDMNVK FFNSNKKKRD DFEKLTNYSV TDLNVQRKAI HELIQVMAEL SPAAKTGKRK RSQMLFRG
Greater than 90% as determined by SDS-PAGE.
3458
50 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers