missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IFN-alpha I (alpha 17) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 111501
Additional Details : Weight : 0.01000kg
Description
The Recombinant Human IFN-alpha I (alpha 17) Protein is derived from E. coli. The Recombinant Human IFN-alpha I (alpha 17) Protein has been validated for the following applications: Bioactivity.Specifications
V00532 | |
Phosphate buffered saline (PBS) containing 0.1% bovine serum albumin (BSA) | |
E. coli-derived human IFN-alpha I protein | |
Unconjugated |
Bioactivity | |
Predicted MW: 19.3kDa | |
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |