missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IFIH1 Protein
A DNA sequence encoding the Homo sapiens (Human) IFIH1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8166-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_071451.2 | |
IFIH1 Protein | |
Research Use Only | |
IFIH1 | |
Recombinant Protein | |
E. coli | |
KLTKLRNTIM EQYTRTEESA RGIIFTKTRQ SAYALSQWIT ENEKFAEVGV KAHHLIGAGH SSEFKPMTQN EQKEVISKFR TGKINLLIAT TVAEEGLDIK ECNIVIRYGL VTNEIAMVQA RGRARADEST YVLVAHSGSG VIEHETVNDF REKMMYKAIH CVQNMKPEEY AHKILELQMQ SIMEKKMKTK RNIAKHYKNN PSLITFLCKN CSVLACSGED IHVIEKMHHV NMTPEFKELY IVRENKALQK KCADYQINGE IICKCGQAWG TMMVHKGLDL PCLKIRNFVV VFKNNSTKKQ YKKWVELPIT FPNLDYSECC LFSDED | |
Greater than 80% as determined by SDS-PAGE. |
64135 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |