missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IDH3B Protein
A DNA sequence encoding the Homo sapiens (Human) IDH3B, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_001317692.1 |
---|---|
Gene ID (Entrez) | 3420 |
Name | IDH3B Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16011460
|
enQuireBio™
QP6193-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
16031460
|
enQuireBio™
QP6193-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
16021460
|
enQuireBio™
QP6193-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16041460
|
enQuireBio™
QP6193-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16051460
|
enQuireBio™
QP6193-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16061460
|
enQuireBio™
QP6193-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001317692.1 | |
IDH3B Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
3420 | |
Research Use Only | |
IDH3B | |
Recombinant Protein | |
E. coli | |
ASRSQAEDVR VEGSFPVTML PGDGVGPELM HAVKEVFKAA AVPVEFQEHH LSEVQNMASE EKLEQVLSSM KENKVAIIGK IHTPMEYKGE LASYDMRLRR KLDLFANVVH VKSLPGYMTR HNNLDLVIIR EQTEGEYSSL EHESARGVIE CLKIVTRAKS QRIAKFAFDY ATKKGRGKVT AVHKANIMKL GDGLFLQCCE EVAELYPKIK FETMIIDNCC MQLVQNPYQF DVLVMPNLYG NIIDNLAAGL VGGAGVVPGE SYSAEYAVFE TGARHPFAQA VGRNIANPTA MLLSASNMLR HLNLEYHSSM IADAVKKVIK VGKVRTRDMG GYSTTTDFIK SVIGHLQTKG S | |
Greater than 90% as determined by SDS-PAGE. |